Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QNEGCONSET

QuicTPD Negative Control Set

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

packaging

pkg of 10 mg

Quality Level

storage temp.

2-8°C

General description

N-Methylated Pomalidomide is a ligand used as a negative control for pomalidomide in the recuitment of Cereblon (CRBN) protein, whereas (S,S,S) AHPC-hydrochloride is a ligand used as negative control for (S,R,S)-AHPC in the recruitment of von-Hippel Lindau (VHL) protein. N-methylation of pomalidomide or the inversion of stereochemistry to yield of (S,S,S) AHPC prevents binding to CRBN and VHL proteins. These are provided at 10mg each.

Application

Useful for construction of small molecules structurally similar to PROTACs of interest but with no inherent degradation action in themselves serving as negative control.

Features and Benefits

Structural similarity to active CRBN/VHL ligands is optimal for construction of negative controls. The amino terminus in each can be used for attaching bifunctional linkers with reactions such as peptide coupling.

Other Notes

Negative Control Set part of QuicTPD kit

Legal Information

QuicTPD is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service