Skip to Content
MilliporeSigma

QTPDAMINE

QuicTPD Amine Screening Set

greener alternative

Greener Alternative

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.21
UNSPSC Code:
41116100
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

General description

QTPD Amine set comprised of 36 partial protacs plated in duplicate with rich linker diversity, architecture and length targeting Cereblon(CRBN) and von Hippel Lindau( VHL) E3 ligases ending in a amine terminal.
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of the four categories of greener alternatives. Click here to view its DfS scorecard.

Application

This set can be used for reacting with warheads/ligands for targets of interest with solvent exposed pendant carboxylic acid handles via peptide coupling reactions. Two distinct warheads can be reacted with this set to generate 72 distinct small molecule degraders that can be used for subsequent screening.

Features and Benefits

Representative set of partial protacs targeting both CRBN and VHL with typical PEG, hydrophobic and rigid linkers that can be used for construction of PROTACs. Customers can make screen upto 24 reaction conditions with this set and empty glass shell vials provided with the pre-plated set. Glass shell vials allow for exploration of reaction conditions tolerant to different, reagents, solvents and temperature. Miniaturized synthesis allows for reduction in solvent, reagent, water and energy usage making this a sustainable and faster route to PROTAC discovery.

Other Notes

QuicTPD Amine Screening Set

PROTAC Coupling Model Calculations available under the More Documents section below.

Legal Information

QuicTPD is a trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

target_organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Health hazardExclamation mark

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library