MilliporeSigma
All Photos(1)

T10006

Sigma-Aldrich

Myristaldehyde

technical grade, 80%

Synonym(s):
Myristic aldehyde, Tetradecanal, Tetradecyl aldehyde
Linear Formula:
CH3(CH2)12CHO
CAS Number:
Molecular Weight:
212.37
Beilstein:
1765987
EC Number:
MDL number:

grade

technical grade

assay

80%

bp

166 °C/24 mmHg (lit.)

SMILES string

CCCCCCCCCCCCCC=O

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service