MilliporeSigma
All Photos(1)

T8175

Sigma-Aldrich

(3aS,4R,6S,7aS)-Tetrahydro-4-hydroxymethyl-6-methoxy-4H-furo[3,2-c]pyran-2(3H)-one

≥98%

Synonym(s):
Methyl 2-deoxy-α-D-alloside 3,4-carbonate
Empirical Formula (Hill Notation):
C9H14O5
CAS Number:
Molecular Weight:
202.20
MDL number:
PubChem Substance ID:

assay

≥98%

storage temp.

−20°C

SMILES string

CO[C@H]1C[C@H]2OC(=O)C[C@@H]2[C@H](CO)O1

InChI

1S/C9H14O5/c1-12-9-3-6-5(2-8(11)13-6)7(4-10)14-9/h5-7,9-10H,2-4H2,1H3/t5-,6+,7-,9+/m0/s1

InChI key

CEVALZNMSHYICJ-VOQBNFLRSA-N

Biochem/physiol Actions

Precursor in synthesis of thromboxane derivatives.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service