All Photos(1)

W215007

Sigma-Aldrich

Benzyl propionate

≥98%, FCC

Linear Formula:
C2H5CO2CH2C6H5
CAS Number:
Molecular Weight:
164.20
FEMA Number:
2150
EC Number:
Council of Europe no.:
413
MDL number:
PubChem Substance ID:
Flavis number:
9.132

biological source

synthetic

grade

Kosher

reg. compliance

FCC
FDA 21 CFR 172.515

assay

≥98%

refractive index

n20/D 1.497 (lit.)

bp

222 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

Organoleptic

apple; banana; berry; jam; jasmine; strawberry; sweet

application(s)

flavors and fragrances

food allergen

no known allergens

SMILES string

CCC(=O)OCc1ccccc1

InChI

1S/C10H12O2/c1-2-10(11)12-8-9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

VHOMAPWVLKRQAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Taste at 30 ppm

Certificate of Analysis

Certificate of Origin

Mosciano, G.
Perfumer & Flavorist, 24, 49-49 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service