MilliporeSigma
All Photos(2)

W231401

Sigma-Aldrich

Citronellyl formate

FCC

Linear Formula:
HCO2CH2CH2CH(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
184.28
FEMA Number:
2314
EC Number:
Council of Europe no.:
345
MDL number:
PubChem Substance ID:
Flavis number:
9.078
NACRES:
NA.21

Quality Level

biological source

synthetic

grade

Kosher

reg. compliance

FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

refractive index

n20/D 1.446 (lit.)

bp

235 °C (lit.)

density

0.897 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apricot; floral; peach; plum; rose

SMILES string

CC(CCOC=O)CC\C=C(\C)C

InChI

1S/C11H20O2/c1-10(2)5-4-6-11(3)7-8-13-9-12/h5,9,11H,4,6-8H2,1-3H3

InChI key

DZNVIZQPWLDQHI-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

H J Fang et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 24(5), 366-371 (1989-01-01)
Three geranium oils were separated, identified and compared by high resolution cross-linked fused silica capillary GC and GC-MSD. It was found that their chemical constituents are very similar, but the contents of some components (linalool, citronellol, cis-caryophyllene and guaiene) differ
Zuobing Xiao et al.
Food research international (Ottawa, Ont.), 116, 211-222 (2019-02-06)
This study focused on the impact of esters on the perception of floral aroma in rose essential oil. Various aromatic reconstitutions were prepared, consisting of 10 alcohols and 9 esters, all the concentrations found in rose essential oil. Sensory analysis
Mohamed Nadjib Boukhatem et al.
The Libyan journal of medicine, 8, 22520-22520 (2013-10-10)
Since the available anti-inflammatory drugs exert an extensive variety of side effects, the search for new anti-inflammatory agents has been a priority of pharmaceutical industries. The aim of the present study was to assess the anti-inflammatory activities of the essential
A Gallardo et al.
Journal of medical entomology, 49(2), 332-335 (2012-04-13)
The major components of geranium (Geranium maculatum L.) oil and their mixtures were tested against female Pediculus humanus capitis De Geer (Phthiraptera: Pediculidae). Chemical analysis by gas chromatography coupled to mass spectrometry revealed four major constituents: citronellol (38%), geraniol (16%)

Protocols

GC Analysis of Geranium Bourbon Essential Oil on SUPELCOWAX® 10

-(+)-Limonene, purum, ≥98.0% (sum of enantiomers, GC); Geranyl tiglate; α-Terpineol, natural, ≥96%, FCC, FG; Geranyl formate; α-Pinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service