W239901

Sigma-Aldrich

Sodium phosphate dibasic

FG

Synonym(s):
Sodium hydrogenphosphate, Disodium phosphate, Disodium hydrogen phosphate, sec-Sodium phosphate
Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
FEMA Number:
2398
EC Number:
MDL number:
PubChem Substance ID:
E Number:
E 339

Quality Level

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1333/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.1778
FDA 21 CFR 182.6290

form

powder

pH

8.9-9.2 (25 °C, 50 g/L in water)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

solubility

water: soluble

Documentation

see Safety & Documentation for available documents

Featured Industry

Flavors and Fragrances

Organoleptic

odorless

food allergen

no known allergens

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Samuel Adler et al.
Gastrointestinal endoscopy, 76(6), 1170-1174 (2012-10-03)
Colon capsule endoscopy (CCE) is a noninvasive technique for the detection of colorectal lesions. However, for CCE to be offered as an out-of-clinic procedure, the system needs to automatically alert the patient when to ingest the laxative (booster). We tested...
Megan A Rees et al.
Journal of proteome research, 14(1), 120-132 (2014-10-21)
Interactions between a host and a bacterial pathogen are mediated by cross-talk between molecules present on, or secreted by, pathogens and host binding-molecules. Identifying proteins involved at this interface would provide substantial insights into this interaction. Although numerous studies have...
Natalie K Binder et al.
Molecular human reproduction, 21(5), 424-434 (2015-03-04)
The rate of obesity among men of reproductive age has tripled in the last three decades. Previously, we demonstrated that paternal obesity resulted in impaired preimplantation developmental kinetics, compromised post-compaction metabolism and decreased blastocyst cell number when embryos were generated...
Johanna Troberg et al.
Drug metabolism and disposition: the biological fate of chemicals, 43(1), 107-118 (2014-10-11)
Understanding drug glucuronidation in the dog, a preclinical animal, is important but currently poorly characterized at the level of individual enzymes. We have constructed cDNAs for the 10 dog UDP-glucuronosyltransferases of subfamily 1A (dUGT1As), expressed them in insect cells, and...
Jiwei Cui et al.
ACS nano, 9(2), 1571-1580 (2015-02-26)
We report the engineering of poly(ethylene glycol) (PEG) hydrogel particles using a mesoporous silica (MS) templating method via tuning the PEG molecular weight, particle size, and the presence or absence of the template and investigate the cell association and biodistribution...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.