Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W255009

Sigma-Aldrich

Heptyl isobutyrate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2(CH2)6CH3
CAS Number:
Molecular Weight:
186.29
FEMA Number:
2550
EC Number:
Council of Europe no.:
295
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.420
Pricing and availability is not currently available.

biological source

synthetic

reg. compliance

FDA 21 CFR 172.515

assay

≥98%

refractive index

n20/D 1.418 (lit.)

bp

212 °C (lit.)

density

0.863 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

organoleptic

apricot; cherry; grape; green; oily; orange; herbaceous; woody; peach; pear; pineapple; plum; raspberry; wine-like

SMILES string

CCCCCCCOC(=O)C(C)C

InChI

1S/C11H22O2/c1-4-5-6-7-8-9-13-11(12)10(2)3/h10H,4-9H2,1-3H3

InChI key

RFDUMBPGZUIKOG-UHFFFAOYSA-N

Biochem/physiol Actions

Odor at 1%
Taste at 5-20 ppm

Other Notes

Natural occurrence: Hop Oil.
Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

140.0 °F - closed cup

flash_point_c

60 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 26, 80-80 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service