All Photos(1)

W301906

Sigma-Aldrich

Skatole

≥98%

Synonym(s):
3-Methylindole, Skatole
Empirical Formula (Hill Notation):
C9H9N
CAS Number:
Molecular Weight:
131.17
FEMA Number:
3019
Beilstein:
111296
EC Number:
Council of Europe no.:
493
MDL number:
PubChem Substance ID:
Flavis number:
14.004

Quality Level

biological source

synthetic

grade

Halal
Kosher

assay

≥98%

bp

265-266 °C (lit.)

mp

92-97 °C (lit.)

Documentation

see Safety & Documentation for available documents

Featured Industry

Flavors and Fragrances

food allergen

no known allergens

SMILES string

Cc1c[nH]c2ccccc12

InChI

1S/C9H9N/c1-7-6-10-9-5-3-2-4-8(7)9/h2-6,10H,1H3

InChI key

ZFRKQXVRDFCRJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in glass bottles

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Certificate of Analysis

Certificate of Origin

Walter S Leal et al.
PloS one, 3(8), e3045-e3045 (2008-08-30)
Synthetic mosquito oviposition attractants are sorely needed for surveillance and control programs for Culex species, which are major vectors of pathogens causing various human diseases, including filariasis, encephalitis, and West Nile encephalomyelitis. We employed novel and conventional chemical ecology approaches...
Mechanisms of 3-methylindole pneumotoxicity.
G S Yost
Chemical research in toxicology, 2(5), 273-279 (1989-09-01)
B Marcos et al.
Meat science, 95(3), 688-693 (2012-11-28)
Expression of water soluble proteins of fresh pork Longissimus thoracis from 4 pure breed pigs (Duroc, Large White, Landrace, and Piétrain) was studied to identify candidate protein markers for meat quality. Surface-enhanced laser desorption/ionisation time-of-flight mass spectrometry (SELDI-TOF-MS) was used...
Hang Wang et al.
Waste management & research : the journal of the International Solid Wastes and Public Cleansing Association, ISWA, 31(2), 223-231 (2013-01-12)
Manure produced from confined animal farms can threaten public and environmental health if not managed properly. Herein, a full-scale commercial bioconversion operation in DeQing County, China for value-added swine manure reduction using house fly, Musca domestica L., larvae is reported....
P K Mathur et al.
Meat science, 91(4), 414-422 (2012-03-23)
A system for sensory evaluation of boar taint was used to evaluate boar taint in fat samples from 6574 entire males. The term "human nose scoring" has been used to describe this system. The samples from each boar were heated...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service