Skip to Content
MilliporeSigma

W307718

p-Tolyl phenylacetate

≥99%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5CH2CO2C6H4CH3
CAS Number:
Molecular Weight:
226.27
FEMA Number:
3077
Council of Europe no.:
236
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.709
EC Number:
202-990-1
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 117

assay

≥99%

mp

74-76 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

hyacinth

SMILES string

Cc1ccc(OC(=O)Cc2ccccc2)cc1

InChI

1S/C15H14O2/c1-12-7-9-14(10-8-12)17-15(16)11-13-5-3-2-4-6-13/h2-10H,11H2,1H3

InChI key

OJEQSSJFSNLMLB-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library