MilliporeSigma
All Photos(2)

W314220

Sigma-Aldrich

3,7-Dimethyl-6-octenoic acid

natural, 97%, FG

Synonym(s):
Citronellic acid, 3,7-Dimethyl-6-octenoic acid
Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CO2H
CAS Number:
Molecular Weight:
170.25
FEMA Number:
3142
EC Number:
MDL number:
PubChem Substance ID:
Flavis number:
8.036
NACRES:
NA.21

Quality Level

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 110

assay

97%

refractive index

n20/D 1.450
n20/D 1.454 (lit.)

bp

121-122 °C/1 mmHg (lit.)

density

0.923 g/mL at 25 °C (lit.)
0.923 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; grassy; green; smoky; floral

SMILES string

CC(CC\C=C(\C)C)CC(O)=O

InChI

1S/C10H18O2/c1-8(2)5-4-6-9(3)7-10(11)12/h5,9H,4,6-7H2,1-3H3,(H,11,12)

InChI key

GJWSUKYXUMVMGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

500 g in poly bottle
1 kg in poly bottle
5 kg in poly drum

Other Notes

Natural occurrence: citronalla java, camphore oil, cypress oil, geranium, lemongrass, mandarin peel, orange leaf, petitgrain combava oil.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service