All Photos(1)

W332305

Sigma-Aldrich

2-Thienyl disulfide

≥95%

Synonym(s):
Dithienyl disulfide
Empirical Formula (Hill Notation):
C8H6S4
CAS Number:
Molecular Weight:
230.39
FEMA Number:
3323
Council of Europe no.:
2333
MDL number:
PubChem Substance ID:
Flavis number:
15.008

biological source

synthetic

grade

Halal
Kosher

assay

≥95%

mp

55-60 °C (lit.)

Documentation

see Safety & Documentation for available documents

Organoleptic

earthy

application(s)

flavors and fragrances

food allergen

no known allergens

SMILES string

S(Sc1cccs1)c2cccs2

InChI

1S/C8H6S4/c1-3-7(9-5-1)11-12-8-4-2-6-10-8/h1-6H

InChI key

YOLFWWMPGNMXFI-UHFFFAOYSA-N

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service