All Photos(1)

W351202

Sigma-Aldrich

2-Methyltetrahydrothiophen-3-one

≥97%

Empirical Formula (Hill Notation):
C5H8OS
CAS Number:
Molecular Weight:
116.18
FEMA Number:
3512
EC Number:
Council of Europe no.:
11601
MDL number:
PubChem Substance ID:
Flavis number:
15.023

biological source

synthetic

grade

Kosher

assay

≥97%

refractive index

n20/D 1.508 (lit.)

bp

82 °C/28 mmHg (lit.)

density

1.119 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

Organoleptic

butter; balsamic

application(s)

flavors and fragrances

food allergen

no known allergens

SMILES string

CC1SCCC1=O

InChI

1S/C5H8OS/c1-4-5(6)2-3-7-4/h4H,2-3H2,1H3

InChI key

YMZZPMVKABUEBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Pascal Fuchsmann et al.
Journal of agricultural and food chemistry, 63(34), 7511-7521 (2015-08-01)
To establish the odor profiles of three differently fabricated commercial Swiss Tilsit cheeses, analyses were conducted using headspace solid-phase microextraction gas chromatography-mass spectrometry/pulsed flame photometric detection and gas chromatography-olfactometry to identify and quantitate volatile compounds. In addition, odor quality and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service