All Photos(1)

W381705

Sigma-Aldrich

2-Acetyl-2-thiazoline

≥96%

Empirical Formula (Hill Notation):
C5H7NOS
CAS Number:
Molecular Weight:
129.18
FEMA Number:
3817
Beilstein:
1209707
Council of Europe no.:
2335
MDL number:
PubChem Substance ID:
Flavis number:
15.01

biological source

synthetic

assay

≥96%

impurities

1% acetic acid

refractive index

n20/D 1.5305 (lit.)

mp

26-30 °C (lit.)

density

1.17 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

application(s)

flavors and fragrances

food allergen

no known allergens

SMILES string

CC(=O)C1=NCCS1

InChI

1S/C5H7NOS/c1-4(7)5-6-2-3-8-5/h2-3H2,1H3

InChI key

FZOZFDAMVVEZSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in glass bottles

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Certificate of Analysis

Certificate of Origin

Chemistry of 2-acetyl-1-pyrroline, 6-acetyl-1,2,3,4-tetrahydropyridine, 2-acetyl-2-thiazoline, and 5-acetyl-2,3-dihydro-4H-thiazine: extraordinary Maillard flavor compounds.
An Adams et al.
Chemical reviews, 106(6), 2299-2319 (2006-06-15)
Kenji Kumazawa et al.
Journal of agricultural and food chemistry, 50(20), 5660-5663 (2002-09-19)
Two kinds of pan-fired green teas (Japanese Kamairi-cha and Chinese Longing tea) were compared with the common Japanese green tea (Sen-cha). Application of the aroma extract dilution analysis (AEDA) using the volatile fraction of the Sen-cha, Kamairi-cha and Longing tea
Rachid Bel Rhlid et al.
Journal of agricultural and food chemistry, 50(8), 2350-2355 (2002-04-04)
Roasted notes contribute to the flavor of thermally processed foods such as meat and bread. 2-Acetyl-2-thiazoline is one of the key volatile compounds responsible for the roasted and popcorn-like aroma character. We report here on the biogeneration of flavoring preparations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service