All Photos(2)

W393713

Sigma-Aldrich

4-Hydroxy-4-methyl-7-cis-decenoic acid γ-lactone

mixture of isomers, ≥97%, FG

Synonym(s):
Waxy lactone, 5-(cis-3-Hexenyl)dihydro-5-methyl-2(3H)furanone, Lactone of cis-jasmone, 4-Methyl-cis-7-decene γ-lactone
Empirical Formula (Hill Notation):
C11H18O2
CAS Number:
Molecular Weight:
182.26
FEMA Number:
3937
EC Number:
Council of Europe no.:
10061
MDL number:
PubChem Substance ID:
Flavis number:
10.061
NACRES:
NA.21

Quality Level

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

description

≥95% cis isomer

assay

≥97%

may contain

<0.10% alpha-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.466

density

0.966 g/mL at 25 °C

Documentation

see Safety & Documentation for available documents

Organoleptic

caramel; coconut; creamy; dairy; oily; fruity; brown; rich

application(s)

flavors and fragrances

food allergen

no known allergens

SMILES string

CC\C=C/CCC1(C)CCC(=O)O1

InChI

1S/C11H18O2/c1-3-4-5-6-8-11(2)9-7-10(12)13-11/h4-5H,3,6-9H2,1-2H3/b5-4-

InChI key

NIKDJTTZUYNCMM-PLNGDYQASA-N

Application

The rich, creamy, oily notes of this lactone will deliver depth and aftertaste to dairy flavors like milk, cream, butter and fresh cheeses. Those same qualities will enhance the fatty acids in nut flavors like pecan, macadamia, Brazil nut and coconut. Sweet brown flavors will also benefit, especially toffee, caramel, dulce de leche, custard, vanilla, butterscotch and chocolate. On the fruit side, it can be used to expand the lactonic profile of peach, apricot, strawberry and mango.

Packaging

100 g in poly bottle
1 kg in poly bottle

Biochem/physiol Actions

Taste at 5-10ppm
Odor at 1%

Certificate of Analysis

Certificate of Origin

Organoleptic Characteristics of Flavor Materials
Michalski, J.
Perfumer & Flavorist, 39(1), 51-51 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service