All Photos(1)

W394440

Sigma-Aldrich

m-Anisic acid

≥98%

Synonym(s):
m-Methylsalicylic acid, m-Anisic acid, 3-Methoxybenzoic acid
Linear Formula:
CH3OC6H4CO2H
CAS Number:
Molecular Weight:
152.15
FEMA Number:
3944
Beilstein:
508838
EC Number:
MDL number:
PubChem Substance ID:
Flavis number:
8.092

biological source

synthetic

assay

≥98%

bp

170-172 °C/10 mmHg (lit.)

mp

105-107 °C (lit.)

Featured Industry

Flavors and Fragrances

Organoleptic

odorless

SMILES string

COc1cccc(c1)C(O)=O

InChI

1S/C8H8O3/c1-11-7-4-2-3-6(5-7)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

XHQZJYCNDZAGLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Certificate of Analysis

Certificate of Origin

A S Waldman et al.
Nucleic acids research, 19(21), 5943-5947 (1991-11-11)
We determined the effect of 3-methoxybenzamide (3-MB), a competitive inhibitor of poly(ADP-ribose) polymerase (E.C. 2.4.2.30), on intrachromosomal homologous recombination in mouse Ltk- cells. We used a cell line that contained in its genome two defective Herpes thymidine kinase (tk) genes...
Thi-Huu Nguyen et al.
Organic letters, 7(12), 2445-2448 (2005-06-04)
[reaction: see text] If employed in THF at 0 degrees C, LTMP metalates meta-anisic acid at the doubly activated position. In contrast, n-BuLi/t-BuOK deprotonates position C-4 preferentially at low temperature. Functionalization at C-6 requires protection of the C-2 site beforehand....
D H Lee et al.
Xenobiotica; the fate of foreign compounds in biological systems, 29(9), 909-916 (1999-11-05)
1. 2-(Allylthio)pyrazine (2-AP) has been demonstrated to protect the liver against toxicants by inhibiting CYP2E1 activity. Since 2-mercaptopyrazine (2-MP) is presumed to be a metabolite of 2-AP, the experiments were performed to determine whether rat liver microsomal and/or cytosolic preparations...
N Dodoff et al.
Journal of inorganic biochemistry, 54(3), 221-233 (1994-05-15)
The complexes [Pt(bah)2X2], [Pt(NH3)(bah)Cl2].0.5H2O, [Pt(mbah)2X2], and [Pt(NH3)(mbah)Cl2] (bah = benzoic acid hydrazide, mbah = 3-methoxybenzoic acid hydrazide; X = Cl, Br, I) have been prepared and characterized by elemental analysis, electric conductivity, IR, 1H NMR, and electronic spectra. A cis-square...
K A DeWeerd et al.
Applied and environmental microbiology, 54(5), 1237-1242 (1988-05-01)
O-methyl substituents of aromatic compounds can provide C1 growth substrates for facultative and strict anaerobic bacteria isolated from diverse environments. The mechanism of the bioconversion of methoxylated benzoic acids to the hydroxylated derivatives was investigated with a model substrate and...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service