W469501

Sigma-Aldrich

Ethyl-2,5-dihydro-4-methylthiazole

≥95%

Synonym(s):
2-Ethyl-4-methyl-2,5-dihydrothiazole
Empirical Formula (Hill Notation):
C6H11NS
CAS Number:
Molecular Weight:
129.22
FEMA Number:
4695
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

assay

≥95%

refractive index

n20/D 1.504

density

1.008 g/mL at 25 °C

Organoleptic

fatty; meaty; nutty; roasted; sulfurous

storage temp.

2-8°C

SMILES string

CC1=NC(CC)SC1

InChI

1S/C6H11NS/c1-3-6-7-5(2)4-8-6/h6H,3-4H2,1-2H3

InChI key

GKTIVNKILASSAW-UHFFFAOYSA-N

Related Categories

General description

Odor Characteristics: 1.0% ETOH. Sulfurous roasted fatty chicken skin, meaty and rubbery with roasted nut nuances of Brazil nut.
Taste Characteristics: @ 0.05 TO 1 PPM. At higher levels, meaty, roasted chicken skin fatty with yeasty and nutty nuances. At lower concentration it imparts interesting and lingering blueberry and peachy fuzzy nuances.

Application

This molecule effects are quite level dependent. It does not really get stronger with higher use levels, it imparts differing characters as you change levels. At higher use levels it is roasted chicken skin and fatty savory with roasted nutty notes while at much lower use levels it imparts lingering fruity blueberry and fuzzy peach notes.

Packaging

1 kg in poly bottle
5 kg in poly drum

RIDADR

UN 3334

WGK Germany

WGK 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.