Documents
Z104116
Supelco
Flash chromatography assembly with standard ball joints
capacity 400 mL, Standard column, Joint: SJ 50/30
NACRES:
NC.05
Joint
Joint: SJ50/30
capacity
400 mL
column O.D. × L
38 mm × 560 mm
General description
Consists of column, flow controller, and pinch clamp. Order solvent reservoir separately.
Application
Flash chromatography is useful for rapid, preparative separations with moderate resolutions. As first described in J. Org. Chem., 43, 2923 (1978), it is commonly performed with silica gel although the use of reversed phase packings has become popular in recent years. For use at <20 psig (1.4 kg/cm2).
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.