Documents

Z192511

Sigma-Aldrich

BD Precisionglide® syringe needles

gauge 20, L 1 in.

Synonym(s):
cannula
NACRES:
NB.03

packaging

pkg of 100 ea

manufacturer/tradename

Becton-Dickinson & Co. 305175

L

1 in.

gauge

20

color

yellow

Looking for similar products? Visit Product Comparison Guide

General description

  • Fits any Luer lock, slip tip, or eccentric tip syringe
  • SS Microlance regular-bevel point
  • Siliconized to reduce resistance
  • PP Luer hub color coded to needle gauge
  • Packaged as individual fiber-free, peel-apart packages in strips of five sterile needles
  • A sheath protects point and cannula
  • All packages carry lot control numbers to insure sterility
  • NoKor needles have scalpel like tip

Legal Information

Precisionglide is a registered trademark of Becton-Dickinson & Co.
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.