Documents
Z236128
Supelco
Ferrule Remover
NACRES:
SB.54
availability
available only in EU
General description
Spiral tapered tip for removal of capillary ferrules from nuts. Two sizes are included, for removing 0.4 mm and 0.8 mm I.D. ferrules.
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.