MilliporeSigma
Z300039
3M disposable half-mask respirator
3M 4254
Select a Size
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
feature
class FFK1P1
manufacturer/tradename
3M 4254
availability
available only in EU
General description
Developed to meet the stringent requirements of the new European Standard EN405, these respirators have one piece carbon filter elements and built-in particulate filters together with a parabolic exhalation valve for easier breathing. A soft, low profile face piece made of special non-allergenic material reduces skin irritation and makes these respirators comfortable to wear.
All these respirators should only be used for protection against hazards with good warning properties (e.g. smell or taste) and providing the concentrations of hazardous substances are not immediately dangerous to life or health (IDLH)
Legal Information
3M is a trademark of 3M Company
Disclaimer
Type of hazard: ammonia/methylamine
Choose from one of the most recent versions:
Certificates of Analysis (COA)