Documents

Z402869

Büchi® beaker flask

distillation, flask working volume 500 mL, Joint: ST/NS 24/40

Pricing and availability is not currently available.

material

flat bottom flask
glass

Joint

Joint: ST/NS24/40

sterility

non-sterile

feature

closure type Standard Taper Joint

packaging

pack of 1

mfr. no.

Büchi, 34247

flask capacity

1,500 mL

flask working volume

500 mL

Looking for similar products? Visit Product Comparison Guide

General description

Two-piece flask offers easy access to product. Flat bottom stands firmly on lab bench. Can be used with bumping or foaming materials or on a lyophilizer to dry sample completely without transfer. Choice of distillation or powder-drying versions.

Legal Information

Büchi is a registered trademark of Büchi Labortechnik AG
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.