All Photos(1)
Z647861
Pyrex® volumetric flask, Class A, works certified
volume 250 mL, precision: 0.15 mL
Joint
joint ST/NS stopper size14/23
volume measuring range
250 mL, precision: 0.15 mL
H
220 mm
General description
Each flask engraved with individual serial number and supplied with certificate of calibration traceable to
National standards.
  • Complies with ISO 1042 and DIN 12664
  • Conformity ′H′ certified
  • Inscriptions in white enamel
  • Polyethylene stopper
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).