Documents
Z740664
Sigma-Aldrich
Aldrich® Essentials Erlenmeyer flask, PTFE lined screw cap
capacity 50 mL, 24-410 thread, volume 20-50 mL, accuracy: 10 mL
NACRES:
NB.43
material
black phenolic cap (with PTFE liner)
borosilicate glass flask
flat bottom
sterility
non-sterile
feature
with cap
packaging
pkg of 24 ea
volume measuring range
20-50 mL, accuracy: 10 mL
capacity
50 mL
fitting
24-410 thread
Related Categories
General description
Clear borosilicate glass Erlenmeyer flask with a threaded top and a black phenolic cap with a PTFE liner. White reference graduations and marking spot. ASTM E1404.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.