MilliporeSigma
All Photos(1)

AMBH2D6EBA00

Ambeed, Inc.

5-Nitro-1H-imidazole

CAS Number:
Molecular Weight:
113.08
MDL number:

bp

404.8�t 760 mmHg

mp

303

Iupac Name

5-nitro-1H-imidazole

Inchi Code

1S/C3H3N3O2/c7-6(8)3-1-4-2-5-3/h1-2H,(H,4,5)

InChI key

VYDWQPKRHOGLPA-UHFFFAOYSA-N

storage temp.

Inert atmosphere,Room Temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/3034-38-6.html

Flash Point

200

Boiling point

404.8�t 760 mmHg

Melting Point

303

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H302-H315-H319-H335

Precautionary statements

MSDS

https://www.ambeed.com/products/3034-38-6.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service