MilliporeSigma
All Photos(1)

AMBH2D6EBE36

Ambeed, Inc.

Z-Trp-OH

CAS Number:
Molecular Weight:
338.36
MDL number:

bp

619.1�t 760 mmHg

mp

121-126

Iupac Name

(2S)-2-{[(benzyloxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid

Inchi Code

1S/C19H18N2O4/c22-18(23)17(10-14-11-20-16-9-5-4-8-15(14)16)21-19(24)25-12-13-6-2-1-3-7-13/h1-9,11,17,20H,10,12H2,(H,21,24)(H,22,23)/t17-/m0/s1

InChI key

AHYFYYVVAXRMKB-KRWDZBQOSA-N

storage temp.

Inert atmosphere,2-8

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/7432-21-5.html

Synonym

(S)-2-(((Benzyloxy)carbonyl)amino)-3-(1H-indol-3-yl)propanoic acid

Flash Point

328.2

Boiling point

619.1�t 760 mmHg

Melting Point

121-126

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/7432-21-5.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service