All Photos(1)

AMBH2D6EC981

Ambeed, Inc.

5- (3,3-Dimethyl-1-triazenyl) imidazole-4-carboxamide

CAS Number:
Molecular Weight:
182.18
MDL number:

Iupac Name

(E)-4-(3,3-dimethyltriaz-1-en-1-yl)-1H-imidazole-5-carboxamide

Inchi Code

1S/C6H10N6O/c1-12(2)11-10-6-4(5(7)13)8-3-9-6/h3H,1-2H3,(H2,7,13)(H,8,9)/b11-10+

InChI key

FDKXTQMXEQVLRF-ZHACJKMWSA-N

storage temp.

Keep in dark place,Sealed in dry,2-8

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/4342-03-4.html

Synonym

4-(3,3-Dimethyltriaz-1-en-1-yl)-1H-imidazole-5-carboxamide

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

H302-H312-H315-H319-H332-H335-H350-H360

Precautionary statements

MSDS

https://www.ambeed.com/products/4342-03-4.html

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service