MilliporeSigma
All Photos(1)

AMBH2D6ECA79

Ambeed, Inc.

Ac-D-Trp-OH

CAS Number:
Molecular Weight:
246.27
MDL number:

bp

586.6�0�t 760 mmHg

mp

180-181

Iupac Name

(2R)-2-(acetylamino)-3-(1H-indol-3-yl)propanoic acid

Inchi Code

1S/C13H14N2O3/c1-8(16)15-12(13(17)18)6-9-7-14-11-5-3-2-4-10(9)11/h2-5,7,12,14H,6H2,1H3,(H,15,16)(H,17,18)/t12-/m1/s1

InChI key

DZTHIGRZJZPRDV-GFCCVEGCSA-N

storage temp.

Keep in dark place,Inert atmosphere,Store in freezer, under -20

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/2280-01-5.html

Synonym

(R)-2-Acetamido-3-(1H-indol-3-yl)propanoic acid

Flash Point

308.6

Boiling point

586.6�0�t 760 mmHg

Melting Point

180-181

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H302-H315-H319-H335

Precautionary statements

MSDS

https://www.ambeed.com/products/2280-01-5.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service