All Photos(1)

AMBH2D6EF95A

Ambeed, Inc.

Tri-m-tolylphosphine

CAS Number:
Molecular Weight:
304.37
MDL number:

bp

423.1�t 760 mmHg

mp

97-99

Iupac Name

tris(3-methylphenyl)phosphine

Inchi Code

1S/C21H21P/c1-16-7-4-10-19(13-16)22(20-11-5-8-17(2)14-20)21-12-6-9-18(3)15-21/h4-15H,1-3H3

InChI key

LFNXCUNDYSYVJY-UHFFFAOYSA-N

storage temp.

Inert atmosphere,Room Temperature

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/6224-63-1.html

Flash Point

221.6

Boiling point

423.1�t 760 mmHg

Melting Point

97-99

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H315-H319-H335

Precautionary statements

MSDS

https://www.ambeed.com/products/6224-63-1.html

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service