Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

AMBH2D6F4271

Ambeed, Inc.

Boc-Asn(Trt)-OH

Synonym(s):

(2S)-2-[(tert-butoxycarbonyl)amino]-4-oxo-4-(tritylamino)butanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C28H30O5N2
CAS Number:
Molecular Weight:
474.56
MDL number:

Inchi Code

1S/C28H30N2O5/c1-27(2,3)35-26(34)29-23(25(32)33)19-24(31)30-28(20-13-7-4-8-14-20,21-15-9-5-10-16-21)22-17-11-6-12-18-22/h4-18,23H,19H2,1-3H3,(H,29,34)(H,30,31)(H,32,33)/t23-/m0/s1

Inchi Key

PYGOCFDOBSXROC-QHCPKHFHSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

solid

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library