Skip to Content
MilliporeSigma

AMBH2D6F4EB2

Ambeed, Inc.

D-(+)-Trehalose dihydrate

Synonym(s):

alpha-D-glucopyranosyl alpha-D-glucopyranoside dihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

10 G

$17.25

25 G

$20.70

100 G

$51.75

500 G

$89.70

1 KG

$177.10

Vendor SKU: A308914-10G

$17.25


Ships in 5 days - from Aldrich Partner.


About This Item

Linear Formula:
C12H26O13
CAS Number:
Molecular Weight:
378.33
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C12H22O11.2H2O/c13-1-3-5(15)7(17)9(19)11(21-3)23-12-10(20)8(18)6(16)4(2-14)22-12;;/h3-20H,1-2H2;2*1H2/t3-,4-,5-,6-,7+,8+,9-,10-,11-,12-;;/m1../s1

Inchi Key

DPVHGFAJLZWDOC-PVXXTIHASA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

solid

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service