MilliporeSigma
All Photos(1)

AMBH2D6F8B62

Ambeed, Inc.

5-Iodo-3-nitropyridin-2-amine

CAS Number:
Molecular Weight:
265.01
MDL number:

bp

365.4�t 760 mmHg

mp

215-219

Iupac Name

5-iodo-3-nitro-2-pyridinamine

Inchi Code

1S/C5H4IN3O2/c6-3-1-4(9(10)11)5(7)8-2-3/h1-2H,(H2,7,8)

InChI key

MDJUWRHSXKZSOJ-UHFFFAOYSA-N

storage temp.

Keep in dark place,Inert atmosphere,2-8

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/25391-57-5.html

Boiling point

365.4�t 760 mmHg

Melting Point

215-219

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/25391-57-5.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service