Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

AMBH2D6FA309

Ambeed, Inc.

Ethyl 3-hydrazinylbenzoate hydrochloride

Synonym(s):

ethyl 3-hydrazinobenzoate hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Product AMBH2D6FA309 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C9H13O2N2Cl1
CAS Number:
Molecular Weight:
216.67
MDL number:

Skip To

Inchi Code

1S/C9H12N2O2.ClH/c1-2-13-9(12)7-4-3-5-8(6-7)11-10;/h3-6,11H,2,10H2,1H3;1H

Inchi Key

YMKGDNLSBACMSE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

solid

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library