Skip to Content
MilliporeSigma

AMBH2D6FBC55

Ambeed, Inc.

Protocatechualdehyde

Synonym(s):

3,4-dihydroxybenzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

5 G

$11.50

10 G

$13.80

25 G

$29.90

100 G

$33.35

500 G

$143.75

1 KG

$244.95

Vendor SKU: A152308-5G

$11.50


Ships in 5 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
138.12
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mp

150-155C

Inchi Code

1S/C7H6O3/c8-4-5-1-2-6(9)7(10)3-5/h1-4,9-10H

Inchi Key

IBGBGRVKPALMCQ-UHFFFAOYSA-N

Storage temp.

Keep in dark place, sealed in dry, room temperature

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Looking for similar products? Visit Product Comparison Guide

MSDS

https://www.ambeed.com/products/139-85-5.html

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service