Skip to Content
MilliporeSigma

AMBH2D6FBE98

Ambeed, Inc.

6-Bromo-1-hexene

Synonym(s):

6-bromo-1-hexene

Sign In to View Organizational & Contract Pricing.

Select a Size

Product AMBH2D6FBE98 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C6H11Br1
CAS Number:
Molecular Weight:
163.06
MDL number:

Skip To

Inchi Code

1S/C6H11Br/c1-2-3-4-5-6-7/h2H,1,3-6H2

Inchi Key

RIMXEJYJXDBLIE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

liquid

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library