AMBH2D6FCA52

Ambeed, Inc.

4-Amino-6-methylpyrimidine-5-carbonitrile

CAS Number:
Molecular Weight:
134.14
MDL number:

Iupac Name

4-amino-6-methyl-5-pyrimidinecarbonitrile

Inchi Code

1S/C6H6N4/c1-4-5(2-7)6(8)10-3-9-4/h3H,1H3,(H2,8,9,10)

InChI key

HFTQOHMFCTUMHL-UHFFFAOYSA-N

storage temp.

Keep in dark place,Inert atmosphere,Room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/76574-44-2.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H302-H319

Precautionary statements

MSDS

https://www.ambeed.com/products/76574-44-2.html

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service