All Photos(1)

AMBH2D6FD827

Ambeed, Inc.

EDTA disodium salt

CAS Number:
Molecular Weight:
336.21
MDL number:

mp

248

Iupac Name

sodium 2,2'-(2-(bis(carboxymethyl)amino)ethylazanediyl)diacetate

Inchi Code

1S/C10H16N2O8.2Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;/q;2*+1/p-2

InChI key

ZGTMUACCHSMWAC-UHFFFAOYSA-L

storage temp.

Inert atmosphere,Room Temperature

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/139-33-3.html

Synonym

Sodium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate

Flash Point

100

Melting Point

248

Signal Word

Danger

Hazard Statements

H303-H332-H373-H412

MSDS

https://www.ambeed.com/products/139-33-3.html

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service