All Photos(1)

AMBH2D6FEFBF

Ambeed, Inc.

Bemegride

CAS Number:
Molecular Weight:
155.2
MDL number:

bp

282�t 760 mmHg

mp

126-129

Iupac Name

4-ethyl-4-methyl-2,6-piperidinedione

Inchi Code

1S/C8H13NO2/c1-3-8(2)4-6(10)9-7(11)5-8/h3-5H2,1-2H3,(H,9,10,11)

InChI key

ORRZGUBHBVWWOP-UHFFFAOYSA-N

storage temp.

Sealed in dry,2-8

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/64-65-3.html

Synonym

4-Ethyl-4-methylpiperidine-2,6-dione

Flash Point

125.8

Boiling point

282�t 760 mmHg

Melting Point

126-129

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H302-H315-H319-H335

Precautionary statements

MSDS

https://www.ambeed.com/products/64-65-3.html

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service