Skip to Content
MilliporeSigma

AMBH2D6FF1FB

Ambeed, Inc.

Ribavirin Related Compound D

Synonym(s):

1H-1,2,4-triazole-5-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C3H4O1N4
CAS Number:
Molecular Weight:
112.09
MDL number:

Inchi Code

1S/C3H4N4O/c4-2(8)3-5-1-6-7-3/h1H,(H2,4,8)(H,5,6,7)

Inchi Key

ZEWJFUNFEABPGL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

solid

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library