Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

AMBH2D700FF1

Ambeed, Inc.

GW4869

Synonym(s):

(2E,2'E)-3,3'-(1,4-phenylene)bis(N-(4-(4,5-dihydro-1H-imidazol-2-yl)phenyl)acrylamide) dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C30H30O2N6Cl2
CAS Number:
Molecular Weight:
577.51
MDL number:

Inchi Code

1S/C30H28N6O2.2ClH/c37-27(35-25-11-7-23(8-12-25)29-31-17-18-32-29)15-5-21-1-2-22(4-3-21)6-16-28(38)36-26-13-9-24(10-14-26)30-33-19-20-34-30;;/h1-16H,17-20H2,(H,31,32)(H,33,34)(H,35,37)(H,36,38);2*1H/b15-5+,16-6+;;

Inchi Key

NSFKAZDTKIKLKT-CLEIDKRQSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

solid

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library