Skip to Content
MilliporeSigma

AMBH2D70119A

Ambeed, Inc.

2-Chloro-3-(trifluoromethyl)phenylboronic acid

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product AMBH2D70119A is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
224.37
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

2-chloro-3-(trifluoromethyl)phenylboronic acid

Inchi Code

1S/C7H5BClF3O2/c9-6-4(7(10,11)12)2-1-3-5(6)8(13)14/h1-3,13-14H

Inchi Key

NEHJDQKHVZYUAA-UHFFFAOYSA-N

storage temp.

Keep in dark place, sealed in dry, room temperature

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/957061-11-9.html

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/957061-11-9.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library