AMBH2D702676

Ambeed, Inc.

4,7-Dimethyl-1,10-phenanthroline

CAS Number:
Molecular Weight:
208.26
MDL number:

bp

397.5�t 760 mmHg

mp

193-195

Iupac Name

4,7-dimethyl[1,10]phenanthroline

Inchi Code

1S/C14H12N2/c1-9-5-7-15-13-11(9)3-4-12-10(2)6-8-16-14(12)13/h3-8H,1-2H3

InChI key

JIVLDFFWTQYGSR-UHFFFAOYSA-N

storage temp.

Sealed in dry,Room Temperature

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/3248-05-3.html

Flash Point

176.9

Boiling point

397.5�t 760 mmHg

Melting Point

193-195

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H302-H315-H319-H335

Precautionary statements

MSDS

https://www.ambeed.com/products/3248-05-3.html

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service