Skip to Content
MilliporeSigma

AMBH303C5844

Ambeed, Inc.

2-Hydrazinyl-4-methylbenzo[d]thiazole

Synonym(s):

(2E)-4-methyl-1,3-benzothiazol-2(3H)-one hydrazone

Sign In to View Organizational & Contract Pricing.

Select a Size

25 G

$17.25

100 G

$46.00

Vendor SKU: A329208-25G

$17.25


Ships in 5 days - from Aldrich Partner.


About This Item

Linear Formula:
C8H9N3S1
CAS Number:
Molecular Weight:
179.25
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C8H9N3S/c1-5-3-2-4-6-7(5)10-8(11-9)12-6/h2-4H,9H2,1H3,(H,10,11)

Inchi Key

DYWNRVWOUASMDT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

solid

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service