Sign In to View Organizational & Contract Pricing.
Select a Size
5 G
$23.00
10 G
$26.45
25 G
$29.90
100 G
$36.80
500 G
$119.60
Vendor SKU: A351387-5G
$23.00
Ships in 5 days - from Aldrich Partner.
About This Item
Skip To
Inchi Code
1S/C3H7BrO/c1-5-3-2-4/h2-3H2,1H3
Inchi Key
YZUPZGFPHUVJKC-UHFFFAOYSA-N
Shipping Temperature
Cold-chain transportation
Purity
97%
Storage temp.
Inert atmosphere, store in freezer, under -20C
Country of Origin
CN
Physical Form
liquid
hstatements
signalword
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service
