Sign In to View Organizational & Contract Pricing.
Select a Size
25 G
$26.45
Vendor SKU: A627482-25G
$26.45
Ships in 5 days - from Aldrich Partner.
About This Item
Skip To
Inchi Code
1S/C7H13Br/c8-6-7-4-2-1-3-5-7/h7H,1-6H2
Inchi Key
UUWSLBWDFJMSFP-UHFFFAOYSA-N
Storage temp.
Inert atmosphere, room temperature
Purity
98%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
liquid
MSDS
https://www.ambeed.com/products/2550-36-9.html
hstatements
pcodes
signalword
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service