AMBH303C6B32

Ambeed, Inc.

DL-Methionine Methylsulfonium Chloride

CAS Number:
Molecular Weight:
199.7
MDL number:

mp

139�t N/A mmHg

Iupac Name

(3-amino-3-carboxypropyl)(dimethyl)sulfonium chloride

Inchi Code

1S/C6H13NO2S.ClH/c1-10(2)4-3-5(7)6(8)9;/h5H,3-4,7H2,1-2H3;1H

InChI key

MYGVPKMVGSXPCQ-UHFFFAOYSA-N

storage temp.

Inert atmosphere,Room Temperature

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/3493-12-7.html

Synonym

(3-Amino-3-carboxypropyl)dimethylsulfonium chloride

Melting Point

139�t N/A mmHg

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H302-H315-H319-H335

Precautionary statements

MSDS

https://www.ambeed.com/products/3493-12-7.html

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service