MilliporeSigma
All Photos(1)

AMBH47A7207F

Ambeed, Inc.

4-Fluorobenzaldehyde oxime

CAS Number:
Molecular Weight:
139.13
MDL number:

bp

194.9�t 760 mmHg

mp

82-85

Iupac Name

4-fluorobenzaldehyde oxime

Inchi Code

1S/C7H6FNO/c8-7-3-1-6(2-4-7)5-9-10/h1-4H,5H2

InChI key

LYUNTQMDKDIIKH-UHFFFAOYSA-N

storage temp.

Sealed in dry,2-8

Purity

98%

Country of Origin

US

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/459-23-4.html

Synonym

N/A

Flash Point

71.7

Boiling point

194.9�t 760 mmHg

Melting Point

82-85

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service