MilliporeSigma
All Photos(1)

AMBH580648A0

Ambeed, Inc.

1,2-Di(thiophen-2-yl)disulfane

CAS Number:
Molecular Weight:
230.4
MDL number:

bp

325.2�t 760 mmHg

mp

55-60�t N/A mmHg

Iupac Name

2-(2-thienyldisulfanyl)thiophene

Inchi Code

1S/C8H6S4/c1-3-7(9-5-1)11-12-8-4-2-6-10-8/h1-6H

InChI key

YOLFWWMPGNMXFI-UHFFFAOYSA-N

storage temp.

Keep in dark place,Inert atmosphere,Room temperature

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/6911-51-9.html

Flash Point

150.5

Boiling point

325.2�t 760 mmHg

Melting Point

55-60�t N/A mmHg

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

H302-H318

Precautionary statements

MSDS

https://www.ambeed.com/products/6911-51-9.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service