Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

AMBH58064984

Ambeed, Inc.

4-Bromothioanisole

Synonym(s):

1-bromo-4-(methylsulfanyl)benzene

Sign In to View Organizational & Contract Pricing.

Select a Size

5 G

$11.50

25 G

$28.75

100 G

$82.80

Vendor SKU: A887360-5G

$11.50


Ships in 5 days - from Aldrich Partner.


About This Item

Linear Formula:
C7H7Br1S1
CAS Number:
Molecular Weight:
203.1
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C7H7BrS/c1-9-7-4-2-6(8)3-5-7/h2-5H,1H3

Inchi Key

YEUYZNNBXLMFCW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

99%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or Semi-solid or liquid

Melting Point

38-40C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service