Skip to Content
MilliporeSigma

AMBH5817679B

Ambeed, Inc.

N,N,N,N-Tetramethyl-1,4-phenylenediamine

Synonym(s):

N~1~,N~1~,N~4~,N~4~-tetramethyl-1,4-benzenediamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available. Contact Technical Service

About This Item

Linear Formula:
C10H16N2
CAS Number:
Molecular Weight:
164.25
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C10H16N2/c1-11(2)9-5-7-10(8-6-9)12(3)4/h5-8H,1-4H3

Inchi Key

CJAOGUFAAWZWNI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Form: solid or semi-solid or liquid

Melting Point

49-51C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service